missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Tubulin beta-3 chain Protein

A DNA sequence encoding the Homo sapiens (Human) Tubulin beta-3 chain, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8460-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024751

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001184110.1
Tubulin beta-3 chain Protein
Research Use Only
TUBB3
Recombinant Protein
E. coli
MREIVHIQAG QCGNQIGAKF WEVISDEHGI DPSGNYVGDS DLQLERISVY YNEASSHKYV PRAILVDLEP GTMDSVRSGA FGHLFRPDNF IFGQSGAGNN WAKGHYTEGA ELVDSVLDVV RKECENCDCL QGFQLTHSLG GGTGSGMGTL LISKVREEYP DRIMNTFSVV PSPKVSDTVV EPYNATLSIH QLVENTDETY CIDNEALYDI
Greater than 90% as determined by SDS-PAGE.
10381
500 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers