missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TSEN15 Protein
A cDNA sequence encoding the TSEN15 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13824-10ug
Additional Details : Weight : 0.01000kg
Specifications
116461 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MEERGDSEPT PGCSGLGPDG VRGFGDGGGA PSWAPEDAWM GTHPKYLEMM ELDIGDATHV YVAFLVYLDL MESKSWHEVN CVGLPELQLI CLVGTEIEGE GLQTVVPTPI TASLSHNRIR EILKASRKLQ GDPDLPMSFT LAIVESDSTI VYYKLTDGFM LPDPQNISLR R | |
Greater than 90.0% as determined by SDS-PAGE. |
TSEN15 Protein | |
Research Use Only | |
TSEN15 | |
Human | |
His | |
The TSEN15 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 20% glycerol and 200mM NaCl. |