missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transcription elongation factor SPT4 Protein
A DNA sequence encoding the Homo sapiens (Human) Transcription elongation factor SPT4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6753-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003159.1 | |
Transcription elongation factor SPT4 Protein | |
Research Use Only | |
SUPT4H1 | |
Recombinant Protein | |
E. coli | |
ALETVPKDLR HLRACLLCSL VKTIDQFEYD GCDNCDAYLQ MKGNREMVYD CTSSSFDGII AMMSPEDSWV SKWQRVSNFK PGVYAVSVTG RLPQGIVREL KSRGVAYKSR DTAIKT | |
Greater than 80% as determined by SDS-PAGE. |
6827 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |