missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TIMM23 Protein
A DNA sequence encoding the Homo sapiens (Human) TIMM23, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6793-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006318.1 | |
TIMM23 Protein | |
Research Use Only | |
TIMM23 | |
Recombinant Protein | |
E. coli | |
MEGGGGSGNK TTGGLAGFFG AGGAGYSHAD LAGVPLTGMN PLSPYLNVDP RYLVQDTDEF ILPTGANKTR GRFELAFFTI GGCCMTGAAF GAMNGLRLGL KETQNMAWSK PRNVQILNMV TRQGALWANT LGSLALLYSA FGVIIEKTRG AEDDLNTVAA GTMTGMLYKC TGGLRGIARG GLTGLTLTSL YALYNNWEHM KGSLLQQSL | |
Greater than 80% as determined by SDS-PAGE. |
100287932 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |