missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TIMM23 Protein

A DNA sequence encoding the Homo sapiens (Human) TIMM23, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6793-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005900

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_006318.1
TIMM23 Protein
Research Use Only
TIMM23
Recombinant Protein
E. coli
MEGGGGSGNK TTGGLAGFFG AGGAGYSHAD LAGVPLTGMN PLSPYLNVDP RYLVQDTDEF ILPTGANKTR GRFELAFFTI GGCCMTGAAF GAMNGLRLGL KETQNMAWSK PRNVQILNMV TRQGALWANT LGSLALLYSA FGVIIEKTRG AEDDLNTVAA GTMTGMLYKC TGGLRGIARG GLTGLTLTSL YALYNNWEHM KGSLLQQSL
Greater than 80% as determined by SDS-PAGE.
100287932
200 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers