Learn More
R&D Systems™ Recombinant Human TIM-4 Fc Chimera Protein
Description
The Recombinant Human TIM-4 Fc Chimera Protein is derived from HEK293. The Recombinant Human TIM-4 Fc Chimera Protein has been validated for the following applications: Bioactivity.
Order Info
The product is shipped at ambient temperature.
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Source | Human embryonic kidney cell,HEK293-derived human TIM-4 protein, (N-terminus) Human TIM-4 (Glu25-Gln314) Accession &Num; Q96H15 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Product Line | R&D Systems |
For Research Use Only