missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Thymosin beta 4 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
Description
Thymosin beta 4 protein is fully biologically active when compared to standard. The biological activity determined by its ability to produce a protective effect against hydrogen peroxide in primary lung fibroblasts is in a concentration range of 0.5 - 10 μg/mL.
Specifications
Specifications
| Accession Number | P62328 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, pH 7.4. |
| Gene ID (Entrez) | 7114 |
| Molecular Weight (g/mol) | 4.9 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of Thymosin beta 4 as determined by LAL method. |
| Gene Alias | PTMB4, T beta-4, TB4, TB4Xbeta 4, X chromosome, THYB4, thymosin beta 4, X-linked, thymosin beta-4 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction