missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TGFB3 Protein
A DNA sequence encoding the Homo sapiens (Human) TGFB3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6783-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001316867.1 | |
TGFB3 Protein | |
Research Use Only | |
TGFB3 | |
Recombinant Protein | |
E. coli | |
LSTCTTLDFG HIKKKRVEAI RGQILSKLRL TSPPEPTVMT HVPYQVLALY NSTRELLEEM HGEREEGCTQ ENTESEYYAK EIHKFDMIQG LAEHNELAVC PKGITSKVFR FNVSSVEKNR TNLFRAEFRV LRVPNPSSKR NEQRIELFQI LRPDEHIAKQ RYIGGKNLPT RGTAEWLSFD VTDTVREWLL RRESNLGLEI SIHCPCHTFQ PNGDILENIH EVMEIKFKGV DNEDDHGRGD LGRLKKQKDH HNPHLILMMI PPHRLDNPGQ GGQRKKR | |
Greater than 90% as determined by SDS-PAGE. |
7043 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |