missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TGFB1-induced anti-apoptotic factor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) TGFB1-induced anti-apoptotic factor 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6790-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004731.2 | |
TGFB1-induced anti-apoptotic factor 1 Protein | |
Research Use Only | |
TIAF1 | |
Recombinant Protein | |
E. coli | |
MSSPSSPFRE QSFLCAAGDA GEESRVQVLK NEVRRGSPVL LGWVEQAYAD KCVCGPSAPP APTPPSLSQR VMCNDLFKVN PFQLQQFRAD PSTASLLLCP GGLDHKLNLR GKAWG | |
Greater than 80% as determined by SDS-PAGE. |
9220 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |