Learn More
R&D Systems™ Recombinant Human SULT2A1 Protein
Description
The Recombinant Human SULT2A1 Protein is derived from E. coli. The Recombinant Human SULT2A1 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Tris, NaCl and EDTA. |
| Gene ID (Entrez) | 6822 |
| Molecular Weight (g/mol) | 35 kDa |
| Name | Cytosolic Sulfotransferase 2A1/SULT2A1 |
| Quantity | 20 μg |
| Source | E. coli-derived human Cytosolic Sulfotransferase 2A1/SULT2A1 protein Ser2-Glu285,with an N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |