missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SPN / CD43 Protein
A DNA sequence encoding the Homo sapiens (Human) SPN / CD43, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6721-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001025459.1 | |
SPN / CD43 Protein | |
Research Use Only | |
SPN | |
Recombinant Protein | |
E. coli | |
STTAVQTPTS GEPLVSTSEP LSSKMYTTSI TSDPKADSTG DQTSALPPST SINEGSPLWT SIGASTGSPL PEPTTYQEVS IKMSSVPQET PHATSHPAVP ITANSLGSHT VTGGTITTNS PETSSRTSGA PVTTAASSLE TSRGTSGPPL TMATVSLETS KGTSGPPVTM ATDSLETSTG TTGPPVTMTT GSLEPSSGAS GPQVSSVKLS TMMSPTTSTN ASTVPFRNPD ENSR | |
Greater than 90% as determined by SDS-PAGE. |
6693 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |