missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Spermidine synthase Protein

A DNA sequence encoding the Homo sapiens (Human) Spermidine synthase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8349-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024161

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_003123.2
Spermidine synthase Protein
Research Use Only
SRM
Recombinant Protein
E. coli
REGWFRETCS LWPGQALSLQ VEQLLHHRRS RYQDILVFRS KTYGNVLVLD GVIQCTERDE FSYQEMIANL PLCSHPNPRK VLIIGGGDGG VLREVVKHPS VESVVQCEID EDVIQVSKKF LPGMAIGYSS SKLTLHVGDG FEFMKQNQDA FDVIITDSSD PMGPAESLFK ESYYQLMKTA LKEDGVLCCQ GECQWLHLDL IKEMRQFCQS LFPVVAYAYC TIPTYPSGQI GFMLCSKNPS TNFQEPVQPL TQQQVAQMQL KYYNSDVHRA AFVLPEFARK ALNDVS
Greater than 90% as determined by SDS-PAGE.
6723
50 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers