missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SPEG / APEG-1 Protein
A DNA sequence encoding the Homo sapiens (Human) SPEG / APEG-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6720-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001166947.1 | |
SPEG / APEG-1 Protein | |
Research Use Only | |
SPEG | |
Recombinant Protein | |
E. coli | |
MQKARGTRGE DAGTRAPPSP GVPPKRAKVG AGGGAPVAVA GAPVFLRPLK NAAVCAGSDV RLRVVVSGTP QPSLRWFRDG QLLPAPAPEP SCLWLRRCGA QDAGVYSCMA QNE | |
Greater than 90% as determined by SDS-PAGE. |
10290 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |