missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Sonic Hedgehog/Shh Protein, High Activity
Description
The Recombinant Human Sonic Hedgehog/Shh Protein, High Activity is derived from HEK293. The Recombinant Human Sonic Hedgehog/Shh Protein, High Activity has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | Q15465 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 6469 |
| Molecular Weight (g/mol) | Observed M.W.: 18-24kDa, reducing conditions, Theoretical M.W.: 20kDa |
| Quantity | 5μg |
| Source | Human embryonic kidney cell,HEK293-derived human Sonic Hedgehog/Shh protein Cys24-Gly197 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
| Gene Alias | HHG1, HHG-1, HLP3, HPE3, MCOPCB5, MCOPCB5sonic hedgehog (Drosophila) homolog, Shh, ShhNC, SMMCI, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS |
| Common Name | Sonic Hedgehog/Shh |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?