missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SNCA / alpha-Synuclein Protein
A DNA sequence encoding the Homo sapiens (Human) SNCA / alpha-Synuclein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6704-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_000336.1 | |
SNCA / alpha-Synuclein Protein | |
Research Use Only | |
SNCA | |
Recombinant Protein | |
Yeast | |
MDVFMKGLSK AKEGVVAAAE KTKQGVAEAA GKTKEGVLYV GSKTKEGVVH GVATVAEKTK EQVTNVGGAV VTGVTAVAQK TVEGAGSIAA ATGFVKKDQL GKNEEGAPQE GILEDMPVDP DNEAYEMPSE EGYQDYEPEA | |
Greater than 90% as determined by SDS-PAGE. |
6622 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |