missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SNAP25 / SUP Protein
A DNA sequence encoding the Homo sapiens (Human) SNAP25 / SUP, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP1354-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001309831.1 | |
SNAP25 / SUP Protein | |
Research Use Only | |
SNAP25 | |
Recombinant Protein | |
E. coli | |
MAEDADMRNE LEEMQRRADQ LADESLESTR RMLQLVEESK DAGIRTLVML DEQGEQLERI EEGMDQINKD MKEAEKNLTD LGKFCGLCVC PCNKLKSSDA YKKAWGNNQD GVVASQPARV VDEREQMAIS GGFIRRVTND ARENEMDENL EQVSGIIGNL RHMALDMGNE IDTQNRQIDR IMEKADSNKT RIDEANQRAT KMLGSG | |
Greater than 90% as determined by SDS-PAGE. |
6616 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |