missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human SBEM His Protein

Product Code. 18167989 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
Unit Size:
0.02mg
0.1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18167989 0.02 mg 0.02mg
18137709 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18167989 Supplier Novus Biologicals™ Supplier No. NBP2518890.02MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 21-90 of Human SBEM The Recombinant Human SBEM Protein is derived from E. coli. The Recombinant Human SBEM Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.25mg/mL
For Use With (Application) Blocking (Adhesion), SDS-PAGE
Formulation Phosphate buffer saline (pH 7.4) containing 10% glycerol
Gene ID (Entrez) 118430
Molecular Weight (g/mol) TMW: 9.3kDa
Quantity 0.02 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias mucin-like 1, mucin-like protein 1, Protein BS106, SBEMSmall breast epithelial mucin
Common Name SBEM
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >85%, by SDS-PAGE
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.