missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S100A2 Protein
A cDNA sequence encoding the S100A2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13392-1mg
Additional Details : Weight : 0.01000kg
Specifications
S100A2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MCSSLEQALA VLVTTFHKYS CQEGDKFKLS KGEMKELLHK ELPSFVGEKV DEEGLKKLMG SLDENSDQQV DFQEYAVFLA LITVMCNDFF QGCPDRP | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The S100A2 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 5mM DTT, 30% glycerol and 0.2M NaCl. |