missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Rhomboid-related protein 1

A DNA sequence encoding the Homo sapiens (Human) Rhomboid-related protein 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6607-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16034510

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_060291.2
Rhomboid-related protein 1
Research Use Only
RHBDL2
Recombinant Protein
E. coli
MNLNMGREMK EELEEEEKMR EDGGGKDRAK SKKVHRIVSK WMLPEKSRGT YLERANCFPP PVFIISISLA ELAVFIYYAV WKPQKQWITL DTGILESPFI YSPEKREEAW RFISYMLVHA G
Greater than 90% as determined by SDS-PAGE.
54933
50 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers