missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Rhomboid-related protein 1
A DNA sequence encoding the Homo sapiens (Human) Rhomboid-related protein 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6607-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_060291.2 | |
Rhomboid-related protein 1 | |
Research Use Only | |
RHBDL2 | |
Recombinant Protein | |
E. coli | |
MNLNMGREMK EELEEEEKMR EDGGGKDRAK SKKVHRIVSK WMLPEKSRGT YLERANCFPP PVFIISISLA ELAVFIYYAV WKPQKQWITL DTGILESPFI YSPEKREEAW RFISYMLVHA G | |
Greater than 90% as determined by SDS-PAGE. |
54933 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |