Learn More
enQuireBio™ Recombinant Human Rho guanine nucleotide exchange factor 18 Protein
A DNA sequence encoding the Homo sapiens (Human) Rho guanine nucleotide exchange factor 18, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8503-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001124427.1 | |
Rho guanine nucleotide exchange factor 18 Protein | |
Research Use Only | |
ARHGEF18 | |
Recombinant Protein | |
E. coli | |
MTVSQKGGPQ PTPSPAGPGT QLGPITGEMD EADSAFLKFK QTADDSLSLT SPNTESIFVE DPYTASLRSE IESDGHEFEA ESWSLAVDAA YAKKQKREVV KRQDVLYELM QTEVHHVRTL KIMLKVYSRA LQEELQFSSK AIGRLFPCAD DLLETHSHFL ARLKERRQES LEEGSDRNYV IQKIGDLLVQ QFSGENGERM KEKYGVFCSG HNEAVSHYKL LLQQNKKFQN LIKKIGNFSI VRRLGVQECI LLVTQRITKY PVLVERIIQN TEAGTEDYED LTQALNLIKD IISQVDAKVS ECEKGQRLRE IAGKMDLKSS SKLKNGLTFR KEDMLQRQLH LEGMLCWKTT SGRLKDILAI LLTDVLLLLQ EKDQKYVFAS VDSKPPVISL QKL | |
Greater than 90% as determined by SDS-PAGE. |
23370 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |