missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Reticulocalbin-1 Protein

A DNA sequence encoding the Homo sapiens (Human) Reticulocalbin-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9756-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16078671

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_002892.1
Reticulocalbin-1 Protein
Research Use Only
RCN1
Recombinant Protein
Yeast
PTVRKERVVR PDSELGERPP EDNQSFQYDH EAFLGKEDSK TFDQLTPDES KERLGKIVDR IDNDGDGFVT TEELKTWIKR VQKRYIFDNV AKVWKDYDRD KDDKISWEEY KQATYGYYLG NPAEFHDSSD HHTFKKMLPR DERRFKAADL NGDLTATREE FTAFLHPEEF EHMKEIVVLE TLEDIDKNGD GFVDQDEYIA DMFSHEENGP EPDWVLSERE QFNEFRDLNK DGKLDKDEIR HWILPQDYDH AQAEARHLVY ESDKNKDEKL TKEEILENWN MFVGSQATNY GEDLTKNHDE L
Greater than 90% as determined by SDS-PAGE.
5954
50 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers