Learn More
enQuireBio™ Recombinant Human RELM-beta Protein
Recombinant Protein
Brand:  enQuireBio™ QP5281-25ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15945839
  • 2650.00 NOK / 25µg
Estimated Shipment: 03-07-2024
to see stock.
Specifications
RELM-beta Protein
Research Use Only
RELM-beta
Human
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
25 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Recombinant Protein
E. coli
MQCSLDSVMD KKIKDVLNSL EYSPSPISKK LSCASVKSQG RPSSCPAGMA VTGCACGYGC GSWDVQLETT CHCQCSVVDW TTARCCHLT
Greater than 90 as determined by SDS-PAGE.