Learn More
R&D Systems™ Recombinant Human Reg3A Protein
Description
The Recombinant Human Reg3A Protein is derived from E. coli. The Recombinant Human Reg3A Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 5068 |
| Molecular Weight (g/mol) | 17 kDa |
| Name | Reg3A |
| Quantity | 50 μg |
| Source | E. coli-derived human Reg3A protein Glu27-Asp175,with an N-terminal Met |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Product Line | R&D Systems |
| Show More |