missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human REG3A / HIP Protein
A DNA sequence encoding the Homo sapiens (Human) REG3A / HIP, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6593-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002571.1 | |
REG3A / HIP Protein | |
Research Use Only | |
REG3A | |
Recombinant Protein | |
E. coli | |
EEPQRELPSA RIRCPKGSKA YGSHCYALFL SPKSWTDADL ACQKRPSGNL VSVLSGAEGS FVSSLVKSIG NSYSYVWIGL HDPTQGTEPN GEGWEWSSSD VMNYFAWERN PSTISSPGHC ASLSRSTAFL RWKDYNCNVR LPYVCKFTD | |
Greater than 90% as determined by SDS-PAGE. |
5068 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |