missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ras-related GTP-binding protein 1
A DNA sequence encoding the Homo sapiens (Human) Ras-related GTP-binding protein A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8446-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006561.1 | |
Ras-related GTP-binding protein 1 | |
Research Use Only | |
RAGA | |
Recombinant Protein | |
E. coli | |
MPNTAMKKKV LLMGKSGSGK TSMRSIIFAN YIARDTRRLG ATIDVEHSHV RFLGNLVLNL WDCGGQDTFM ENYFTSQRDN IFRNVEVLIY VFDVESRELE KDMHYYQSCL EAILQNSPDA KIFCLVHKMD LVQEDQRDLI FKEREEDLRR LSRPLECACF RTSIWDETLY KAWSSIVYQL IPNVQQLEMN LRNFAQIIEA DEVLLFERAT FLVISHYQCK EQRDVHRFEK ISNIIKQFKL SCSKLAA | |
Greater than 90% as determined by SDS-PAGE. |
10670 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |