missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ras-related C3 botulinum toxin substrate 3 Protein

A DNA sequence encoding the Homo sapiens (Human) Ras-related C3 botulinum toxin substrate 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6576-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16034280

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_005043.1
Ras-related C3 botulinum toxin substrate 3 Protein
Research Use Only
RAC3
Recombinant Protein
E. coli
MQAIKCVVVG DGAVGKTCLL ISYTTNAFPG EYIPTVFDNY SANVMVDGKP VNLGLWDTAG QEDYDRLRPL SYPQTDVFLI CFSLVSPASF ENVRAKWYPE VRHHCPHTPI LLVGTKLDLR DDKDTIERLR DKKLAPITYP QGLAMAREIG SVKYLECSAL TQRGLKTVFD EAIRAVLCPP PVKKPGKKC
Greater than 80% as determined by SDS-PAGE.
5881
200 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers