missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ras-related C3 botulinum toxin substrate 3 Protein
A DNA sequence encoding the Homo sapiens (Human) Ras-related C3 botulinum toxin substrate 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6576-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005043.1 | |
Ras-related C3 botulinum toxin substrate 3 Protein | |
Research Use Only | |
RAC3 | |
Recombinant Protein | |
E. coli | |
MQAIKCVVVG DGAVGKTCLL ISYTTNAFPG EYIPTVFDNY SANVMVDGKP VNLGLWDTAG QEDYDRLRPL SYPQTDVFLI CFSLVSPASF ENVRAKWYPE VRHHCPHTPI LLVGTKLDLR DDKDTIERLR DKKLAPITYP QGLAMAREIG SVKYLECSAL TQRGLKTVFD EAIRAVLCPP PVKKPGKKC | |
Greater than 80% as determined by SDS-PAGE. |
5881 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |