missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Rab effector Noc2 Protein

A DNA sequence encoding the Homo sapiens (Human) Rab effector Noc2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8248-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16093541

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001177340.1
Rab effector Noc2 Protein
Research Use Only
RPH3AL
Recombinant Protein
E. coli
MADTIFGSGN DQWVCPNDRQ LALRAKLQTG WSVHTYQTEK QRRKQHLSPA EVEAILQVIQ RAERLDVLEQ QRIGRLVERL ETMRRNVMGN GLSQCLLCGE VLGFLGSSSV FCKDCRKKVC TKCGIEASPG QKRPLWLCKI CSEQREVWKR SGAWFYKGLP KYILPLKTPG RADDPHFRPL PTEPAEREPR SSETSRIYTW ARGRVVSSDS DSDSDLSSSS LEDRLPSTGV RDRKGDKPWK ESGGSVEAPR MGFTHPPGHL SGCQSSLASG ETGTGSADPP GGPRPGLTRR APVKDTPGRA PAADAAPAGP SSCLG
Greater than 90% as determined by SDS-PAGE.
9501
10 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers