missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Rab effector Noc2 Protein
A DNA sequence encoding the Homo sapiens (Human) Rab effector Noc2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8248-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001177340.1 | |
Rab effector Noc2 Protein | |
Research Use Only | |
RPH3AL | |
Recombinant Protein | |
E. coli | |
MADTIFGSGN DQWVCPNDRQ LALRAKLQTG WSVHTYQTEK QRRKQHLSPA EVEAILQVIQ RAERLDVLEQ QRIGRLVERL ETMRRNVMGN GLSQCLLCGE VLGFLGSSSV FCKDCRKKVC TKCGIEASPG QKRPLWLCKI CSEQREVWKR SGAWFYKGLP KYILPLKTPG RADDPHFRPL PTEPAEREPR SSETSRIYTW ARGRVVSSDS DSDSDLSSSS LEDRLPSTGV RDRKGDKPWK ESGGSVEAPR MGFTHPPGHL SGCQSSLASG ETGTGSADPP GGPRPGLTRR APVKDTPGRA PAADAAPAGP SSCLG | |
Greater than 90% as determined by SDS-PAGE. |
9501 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |