missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein SCO2 homolog, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Protein SCO2 homolog, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6652-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001162580.1 | |
Protein SCO2 homolog, mitochondrial Protein | |
Research Use Only | |
SCO2 | |
Recombinant Protein | |
E. coli | |
PAETGGQGQP QGPGLRTRLL ITGLFGAGLG GAWLALRAEK ERLQQQKRTE ALRQAAVGQG DFHLLDHRGR ARCKADFRGQ WVLMYFGFTH CPDICPDELE KLVQVVRQLE AEPGLPPVQP VFITVDPERD DVEAMARYVQ DFHPRLLGLT GSTKQVAQAS HSYRVYYNAG PKDEDQDYIV DHSIAIYLLN PDGLFTDYYG RSRSAEQISD SVRRHMAAFR SVLS | |
Greater than 90% as determined by SDS-PAGE. |
9997 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |