missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein chibby homolog 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein chibby homolog 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8286-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001002880.2 | |
Protein chibby homolog 1 Protein | |
Research Use Only | |
CBY1 | |
Recombinant Protein | |
E. coli | |
MPFFGNTFSP KKTPPRKSAS LSNLHSLDRS TREVELGLEY GSPTMNLAGQ SLKFENGQWI AETGVSGGVD RREVQRLRRR NQQLEEENNL LRLKVDILLD MLSESTAESH LMEKELDELR ISRKRK | |
Greater than 80% as determined by SDS-PAGE. |
25776 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |