missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Proteasome subunit beta type-4 Protein

A DNA sequence encoding the Homo sapiens (Human) Proteasome subunit beta type-4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8363-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16004241

  • 2418.75 NOK / 50µg


Estimated Shipment
24-11-2020
Add to basket

Specifications

Specifications

NP_002787.2
Proteasome subunit beta type-4 Protein
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
5692
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PSMB4
Recombinant Protein
E. coli
TQNPMVTGTS VLGVKFEGGV VIAADMLGSY GSLARFRNIS RIMRVNNSTM LGASGDYADF QYLKQVLGQM VIDEELLGDG HSYSPRAIHS WLTRAMYSRR SKMNPLWNTM VIGGYADGES FLGYVDMLGV AYEAPSLATG YGAYLAQPLL REVLEKQPVL SQTEARDLVE RCMRVLYYRD ARSYNRFQIA TVTEKGVEIE GPLSTETNWD IAHMISGFE
Videos
SDS
Documents