missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Programmed cell death protein 2-like Protein
A DNA sequence encoding the Homo sapiens (Human) Programmed cell death protein 2-like, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8106-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_115722.1 | |
Programmed cell death protein 2-like Protein | |
Research Use Only | |
PDCD2L | |
Recombinant Protein | |
E. coli | |
AAVLKPVLLG LRDAPVHGSP TGPGAWTASK LGGIPDALPT VAAPRPVCQR CGQPLALVVQ VYCPLEGSPF HRLLHVFACA CPGCSTGGAR SWKVFRSQCL QVPEREAQDA QKQGNSLAAE DWCEGADDWG SDTEEGPSPQ FTLDFGNDAS SAKDVDWTAR LQDLRLQDAV LGAAHPVPPG LPLFLPYYIC VADEDDYRDF VNLDHAHSLL RDYQQREGIA MDQLLSQSLP NDGDEKYEKT IIKSGDQTFY KFMKRIAACQ EQILRYSWSG EPLFLTCPTS EVTELPACSQ CGGQRIFEFQ LMPALVSMLK SANLGLSVEF GTILVYTCEK SCWPPNHQTP MEEFCIIQED PDELLFK | |
Greater than 80% as determined by SDS-PAGE. |
84306 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |