missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PRKCA-binding Protein
A DNA sequence encoding the Homo sapiens (Human) PRKCA-binding protein, was expressed in the hosts and tags indicated.
1644.75 NOK - 24050.00 NOK
Specifications
Accession Number | NP_001034672.1 |
---|---|
Gene ID (Entrez) | 9463 |
Name | PRKCA-binding Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16065001
|
enQuireBio™
QP8506-EC-100UG |
100 μg |
5325.00 NOK
100µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16085001
|
enQuireBio™
QP8506-EC-1MG |
1 mg |
24050.00 NOK
1mg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16075001
|
enQuireBio™
QP8506-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16095001
|
enQuireBio™
QP8506-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16005011
|
enQuireBio™
QP8506-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16015011
|
enQuireBio™
QP8506-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001034672.1 | |
PRKCA-binding Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
9463 | |
Research Use Only | |
PICK1 | |
Recombinant Protein | |
E. coli | |
MFADLDYDIE EDKLGIPTVP GKVTLQKDAQ NLIGISIGGG AQYCPCLYIV QVFDNTPAAL DGTVAAGDEI TGVNGRSIKG KTKVEVAKMI QEVKGEVTIH YNKLQADPKQ GMSLDIVLKK VKHRLVENMS SGTADALGLS RAILCNDGLV KRLEELERTA ELYKGMTEHT KNLLRAFYEL SQTHRAFGDV FSVIGVREPQ | |
Greater than 90% as determined by SDS-PAGE. |