missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Platelet glycoprotein Ib alpha chain Protein
A DNA sequence encoding the Homo sapiens (Human) Platelet glycoprotein Ib alpha chain, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6109-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000164.5 | |
Platelet glycoprotein Ib alpha chain Protein | |
Research Use Only | |
GP1BA | |
Recombinant Protein | |
E. coli | |
HPICEVSKVA SHLEVNCDKR NLTALPPDLP KDTTILHLSE NLLYTFSLAT LMPYTRLTQL NLDRCELTKL QVDGTLPVLG TLDLSHNQLQ SLPLLGQTLP ALTVLDVSFN RLTSLPLGAL RGLGELQELY LKGNELKTLP PGLLTPTPKL EKLSLANNNL TELPAGLLNG LENLDTLLLQ ENSLYTIPKG FFGSHLLPFA FLHGNPWLCN CEILYFRRWL QDNAENVYVW KQGVDVKAMT SNVASVQCDN SDKFPVYKYP GKGCPTLGDE GDTDLYDYYP EEDTEGDKVR ATRTVVKFPT KAHTTPWGLF YSWSTASLDS QMPSSLHPTQ ESTKEQTTFP PRWTPNFTLH MESITFSKTP KSTTEPTPSP TTSEPVPEPA PNMTTLEPTP SPTTPEPTSE PAPSPTTPEP TPIPTIATSP TILVSATSLI TPKSTFLTTT KPVSLLESTK KTIPELDQPP KLRGVLQGHL ESSRNDPFLH PDFCCLLPL | |
Greater than 90% as determined by SDS-PAGE. |
2811 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |