missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Nuclear receptor ROR-gamma Protein

A DNA sequence encoding the Homo sapiens (Human) Nuclear receptor ROR-gamma, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6616-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16044580

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001001523.1
Nuclear receptor ROR-gamma Protein
Research Use Only
RORC
Recombinant Protein
E. coli
MDRAPQRQHR ASRELLAAKK THTSQIEVIP CKICGDKSSG IHYGVITCEG CKGFFRRSQR CNAAYSCTRQ QNCPIDRTSR NRCQHCRLQK CLALGMSRDA VKFGRMSKKQ RDSLHAEVQK QLQQRQQQQQ EPVVKTPPAG AQGADTLTYT LGLPDGQLPL GSSPDLPEAS ACPPGLLKAS GSGPSYSNNL AKAGLNGASC HLEYSPERGK AEGRESFYST GSQLTPDRCG LRFEEHRHPG LGELGQGPDS YGSPSFRSTP EAPYASLTEI EHLVQSVCKS YRETCQLRLE DLLRQRSNIF SREEVTGYQR KSMWEMWERC AHHLTEAIQY VVEFAKRLSG FMELCQNDQI VLLKAGAMEV VLVRMCRAYN ADNRTVFFEG KYGGMELFRA LGCSELISSI FDFSHSLSAL HFSEDEIALY TALVLINAHR PGLQEKRKVE QLQYNLELAF HHHLCKTHRQ SILAKLPPKG KLRSLCSQHV ERLQIFQHLH PIVVQAAFPP LYKELFSTET ESPVGLSK
Greater than 90% as determined by SDS-PAGE.
6097
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers