missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nuclear pore complex protein Nup153 Protein
A DNA sequence encoding the Homo sapiens (Human) Nuclear pore complex protein Nup153, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6439-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001265138.1 | |
Nuclear pore complex protein Nup153 Protein | |
Research Use Only | |
NUP153 | |
Recombinant Protein | |
E. coli | |
KAGSSWQCDT CLLQNKVTDN KCIACQAAKL SPRDTAKQTG IETPNKSGKT TLSASGTGFG DKFKPVIGTW DCDTCLVQNK PEAIKCVACE TPKPGTCVKR ALTLTVVSES AETMTASSSS CTVTTGTLGF GDKFKRPIGS WECSVCCVSN NAEDNKCVSC MSEKPGSSVP ASSSSTVPVS LPSGGSLGLE KFKKPEGSWD CELCLVQNKA DSTKCLACES AKPG | |
Greater than 90% as determined by SDS-PAGE. |
9972 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |