missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NRN1 / Neuritin 1 Protein
A DNA sequence encoding the Homo sapiens (Human) NRN1 / Neuritin 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8091-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001265639.1 | |
NRN1 / Neuritin 1 Protein | |
Research Use Only | |
NRN1 | |
Recombinant Protein | |
E. coli | |
AGKCDAVFKG FSDCLLKLGD SMANYPQGLD DKTNIKTVCT YWEDFHSCTV TALTDCQEGA KDMWDKLRKE SKNLNIQGSL FELCGSGNG | |
Greater than 90% as determined by SDS-PAGE. |
51299 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |