missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Non-specific lipid-transfer Protein
A DNA sequence encoding the Homo sapiens (Human) Non-specific lipid-transfer protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6653-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001007099.1 | |
Non-specific lipid-transfer Protein | |
Research Use Only | |
SCP2 | |
Recombinant Protein | |
E. coli | |
MGFPEAASSF RTHQIEAVPT SSASDGFKAN LVFKEIEKKL EEEGEQFVKK IGGIFAFKVK DGPGGKEATW VVDVKNGKGS VLPNSDKKAD CTITMADSDF LALMTGKMNP QSAFFQGKLK ITGNMGLAMK LQNLQLQPGN AKL | |
Greater than 90% as determined by SDS-PAGE. |
6342 | |
50 μg | |
Not determined. | |
Full Length of Isoform SCP2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |