missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NFASC / Neurofascin Protein

A DNA sequence encoding the Homo sapiens (Human) NFASC / Neurofascin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6416-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16093150

  • 5325.00 NOK / 100µg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001005388.2
NFASC / Neurofascin Protein
Research Use Only
NFASC
Recombinant Protein
E. coli
KRSRGGKYPV REKKDVPLGP EDPKEEDGSF DYSDEDNKPL QGSQTSLDGT IKQQESDDSL VDYGEGGEGQ FNEDGSFIGQ YTVKKDKEET EGNESSEATS PVNAIYSLA
Greater than 90% as determined by SDS-PAGE.
23114
100 μg
Not determined.
Cytoplasmic Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers