missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NDUFB7 Protein
A DNA sequence encoding the Homo sapiens (Human) NDUFB7, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8376-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004137.2 | |
NDUFB7 Protein | |
Research Use Only | |
NDUFB7 | |
Recombinant Protein | |
E. coli | |
GAHLVRRYLG DASVEPDPLQ MPTFPPDYGF PERKEREMVA TQQEMMDAQL RLQLRDYCAH HLIRLLKCKR DSFPNFLACK QERHDWDYCE HRDYVMRMKE FERERRLLQR KKRREKKAAE LAKGQGPGEV DPKVAL | |
Greater than 90% as determined by SDS-PAGE. |
4713 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |