missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Napsin A Protein
A DNA sequence encoding the Homo sapiens (Human) Napsin A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6404-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004842.1 | |
Napsin A Protein | |
Research Use Only | |
NAPSA | |
Recombinant Protein | |
E. coli | |
KPIFVPLSNY RDVQYFGEIG LGTPPQNFTV AFDTGSSNLW VPSRRCHFFS VPCWLHHRFD PKASSSFQAN GTKFAIQYGT GRVDGILSED KLTIGGIKGA SVIFGEALWE PSLVFAFAHF DGILGLGFPI LSVEGVRPPM DVLVEQGLLD KPVFSFYLNR DPEEPDGGEL VLGGSDPAHY IPPLTFVPVT VPAYWQIHME RVKVGPGLTL CAKGCAAILD TGTSLITGPT EEIRALHAAI GGIPLLAGEY IILCSEIPKL PAVSFLLGGV WFNLTAHDYV IQTTRNGVRL CLSGFQALDV PPPAGPFWIL GDVFLGTYVA VFDRGDMKSS ARVGLARART RGADLGWGET AQAQFPG | |
Greater than 90% as determined by SDS-PAGE. |
9476 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |