missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NAA10 Protein

A cDNA sequence encoding the NAA10 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12780-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15951519

  • 1155.00 NOK / 5µg
Estimated Shipment: 30-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NAA10 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
NAA10 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 5mM DTT, 10% glycerol and 200mM NaCl.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MNIRNARPED LMNMQHCNLL CLPENYQMKY YFYHGLSWPQ LSYIAEDENG KIVGYVLAKM EEDPDDVPHG HITSLAVKRS HRRLGLAQKL MDQASRAMIE NFNAKYVSLH VRKSNRAALH LYSNTLNFQI SEVEPKYYAD GEDAYAMKRD LTQMADELRR HLELKEKGRH VVLGAIENKV ESKGNSPPSS GEACREEKGL AAEDSGGDSK DLSEVSETTE STDVKDSSEA SDSAS
Videos
SDS
Documents
Certificates
Special Offers

Special Offers