missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Myosin regulatory lc 2, ventric/card muscle isoform Protein
A DNA sequence encoding the Homo sapiens (Human) Myosin regulatory lc 2, ventricular/cardiac muscle isoform, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8680-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000423.2 | |
Myosin regulatory lc 2, ventric/card muscle isoform Protein | |
Research Use Only | |
MYL2 | |
Recombinant Protein | |
E. coli | |
KRAGGANSNV FSMFEQTQIQ EFKEAFTIMD QNRDGFIDKN DLRDTFAALG RVNVKNEEID EMIKEAPGPI NFTVFLTMFG EKLKGADPEE TILNAFKVFD PEGKGVLKAD YVREMLTTQA ERFSKEEVDQ MFAAFPPDVT GNLDYKNLVH IITHGEE | |
Greater than 90% as determined by SDS-PAGE. |
4633 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |