Learn More
enQuireBio™ Recombinant Human Myosin light chain 1 / 3, skeletal muscle isoform Protein
A DNA sequence encoding the Homo sapiens (Human) Myosin light chain 1/3, skeletal muscle isoform, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP6400-ec-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16083040
  • 5325.00 NOK / 100µg
Estimated Shipment: 04-07-2024
Specifications
NP_524144.1
Myosin light chain 1 / 3, skeletal muscle isoform Protein
Research Use Only
MYL1
Recombinant Protein
E. coli
MSFSADQIAE FKEAFLLFDR TGDSKITLSQ VGDVLRALGT NPTNAEVRKV LGNPSNEELN AKKIEFEQFL PMMQAISNNK DQATYEDFVE GLRVFDKEGN GTVMGAELRH VLATLGEKMK EEEVEALMAG QEDSNGCINY EAFVKHIMSI
Greater than 80% as determined by SDS-PAGE.
4632
100 μg
Not determined.
Full Length of Isoform MLC3
Human
GST
Tris-based buffer, 50% glycerol