missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Myosin light chain 1 / 3, skeletal muscle isoform Protein
A DNA sequence encoding the Homo sapiens (Human) Myosin light chain 1/3, skeletal muscle isoform, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6400-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_524144.1 | |
Myosin light chain 1 / 3, skeletal muscle isoform Protein | |
Research Use Only | |
MYL1 | |
Recombinant Protein | |
E. coli | |
MSFSADQIAE FKEAFLLFDR TGDSKITLSQ VGDVLRALGT NPTNAEVRKV LGNPSNEELN AKKIEFEQFL PMMQAISNNK DQATYEDFVE GLRVFDKEGN GTVMGAELRH VLATLGEKMK EEEVEALMAG QEDSNGCINY EAFVKHIMSI | |
Greater than 80% as determined by SDS-PAGE. |
4632 | |
50 μg | |
Not determined. | |
Full Length of Isoform MLC3 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |