missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MTIF3, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) MTIF3, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8051-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16022381

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001159733.1
MTIF3, mitochondrial Protein
Research Use Only
MTIF3
Recombinant Protein
E. coli
TAPAQLSPIA SAPRLSFLIH AKAFSTAEDT QNEGKKTKKN KTAFSNVGRK ISQRVIHLFD EKGNDLGNMH RANVIRLMDE RDLRLVQRNT STEPAEYQLM TGLQILQERQ RLREMEKANP KTGPTLRKEL ILSSNIGQHD LDTKTKQIQQ WIKKKHLVQI TIKKGKNVDV SENEMEEIFH QILQTMPGIA TFSSRPQAVQ GGKALMCVLR AFSKNEEKAY KETQETQERD TLNKDHGNDK ESNVLHQ
Greater than 80% as determined by SDS-PAGE.
219402
500 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers