Learn More
enQuireBio™ Recombinant Human MTIF3, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) MTIF3, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8051-ec-1mg
Additional Details : Weight : 0.01000kg
Specifica
NP_001159733.1 | |
MTIF3, mitochondrial Protein | |
Research Use Only | |
MTIF3 | |
Recombinant Protein | |
E. coli | |
TAPAQLSPIA SAPRLSFLIH AKAFSTAEDT QNEGKKTKKN KTAFSNVGRK ISQRVIHLFD EKGNDLGNMH RANVIRLMDE RDLRLVQRNT STEPAEYQLM TGLQILQERQ RLREMEKANP KTGPTLRKEL ILSSNIGQHD LDTKTKQIQQ WIKKKHLVQI TIKKGKNVDV SENEMEEIFH QILQTMPGIA TFSSRPQAVQ GGKALMCVLR AFSKNEEKAY KETQETQERD TLNKDHGNDK ESNVLHQ | |
Greater than 80% as determined by SDS-PAGE. |
219402 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |