missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human mRNA export factor Protein
A DNA sequence encoding the Homo sapiens (Human) mRNA export factor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6580-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001015885.1 | |
mRNA export factor Protein | |
Research Use Only | |
RAE1 | |
Recombinant Protein | |
E. coli | |
MSLFGTTSGF GTSGTSMFGS ATTDNHNPMK DIEVTSSPDD SIGCLSFSPP TLPGNFLIAG SWANDVRCWE VQDSGQTIPK AQQMHTGPVL DVCWSDDGSK VFTASCDKTA KMWDLSSNQA IQIAQHDAPV KTIHWIKAPN YSCVMTGSWD KTLKFWDTRS SNPMMVLQLP ERCYCADVIY PMAVVATAER GLIVYQLENQ PSEFRRIESP LKHQHRCVAI FKDKQNKPTG FALGSIEGRV AIHYINPPNP AKDNFTFKCH RSNGTNTSAP QDIYAVNGIA FHPVHGTLAT VGSDGRFSFW DKDARTKLKT SEQLDQPISA CCFNHNGNIF AYASSYDWSK GHEFYNPQKK NYIFLRNAAE ELKPRNKK | |
Greater than 80% as determined by SDS-PAGE. |
8480 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |